accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17318-03-5

IUPAC Name: bromo(3-fluorophenyl)magnesium
Synonyms: bromo(3-fluorophenyl)magnesium
Molecular Weight (g/mol): 199.31
Molecular Formula: C6H4BrFMg
InChi Key: VFPDAAQAKDGSHQ-UHFFFAOYSA-M
SMILES: FC1=CC([Mg]Br)=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)