Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Fluorophenylmagnesium Bromide Solution

Catalog No. 55067125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-067-125ML 4 x 25 mL
55-067-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 55-067-125ML Supplier Sigma Aldrich Supplier No. 5506714X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 17318-03-5
Density 1.024 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C6H4BrFMg
Linear Formula FC6H4MgBr
MDL Number MFCD00672007
Quantity 4 x 25 mL
Molecular Weight (g/mol) 199.3 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.