accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 1732-25-8

IUPAC Name: 4-bromo-1,2,3,6,7,8-hexahydropyrene
Synonyms: 4-bromo-1,2,3,6,7,8-hexahydropyrene
Molecular Weight (g/mol): 287.2
Molecular Formula: C16H15Br
InChi Key: SIJTUAANVIVBQW-UHFFFAOYSA-N
SMILES: BrC1=CC2=C3C(CCC2)=CC=C2CCCC1=C32
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results