Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1,2,3,6,7,8-hexahydropyrene 96.0+%, TCI America™
SDP

Catalog No. B51295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51295G 5 g
B51291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51295G Supplier TCI America Supplier No. B51295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1732-25-8
Molecular Formula C16H15Br
Molecular Weight (g/mol) 287.2
MDL Number MFCD01571021
InChI Key SIJTUAANVIVBQW-UHFFFAOYSA-N
PubChem CID 625451
IUPAC Name 4-bromo-1,2,3,6,7,8-hexahydropyrene
SMILES C1CC2=C3C(=CC(=C4C3=C(CCC4)C=C2)Br)C1

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 287.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,2,3,6,7,8-hexahydropyrene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.