accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1732-26-9

IUPAC Name: 4-bromopyrene
Synonyms: 4-bromopyrene
Molecular Weight (g/mol): 281.15
Molecular Formula: C16H9Br
InChi Key: QMHTZTOPYZKQLC-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results