Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromopyrene 95.0+%, TCI America™
SDP

Catalog No. B28075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28075G 5 g
B28071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28075G Supplier TCI America Supplier No. B28075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1732-26-9
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
InChI Key QMHTZTOPYZKQLC-UHFFFAOYSA-N
PubChem CID 3014036
IUPAC Name 4-bromopyrene
SMILES C1=CC2=C3C(=C1)C=C(C4=CC=CC(=C43)C=C2)Br

Specifications

Melting Point 152°C
Color Yellow
Quantity 5 g
Formula Weight 281.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.