CAS RN 17332-11-5
IUPAC Name:
2-bromo-4-methoxyphenol
Synonyms:
2-bromo-4-methoxyphenol
Molecular Weight (g/mol):
203.04
Molecular Formula:
C7H7BrO2
InChi Key:
LTMSUXSPKZRMAB-UHFFFAOYSA-N
SMILES:
COC1=CC=C(O)C(Br)=C1
1
–
1
of
1
results
1
2-Bromo-4-methoxyphenol 98.0+%, TCI America™
CAS: 17332-11-5 Molecular Formula: C7H7BrO2 Molecular Weight (g/mol): 203.04 MDL Number: MFCD02113712 InChI Key: LTMSUXSPKZRMAB-UHFFFAOYSA-N Synonym: 3-Bromo-4-hydroxyanisole PubChem CID: 297382 IUPAC Name: 2-bromo-4-methoxyphenol SMILES: COC1=CC=C(O)C(Br)=C1