CAS RN 17333-84-5
IUPAC Name:
3-chlorobenzene-1-diazonium
Synonyms:
3-chlorobenzenediazonium
Molecular Weight (g/mol):
139.56
Molecular Formula:
C6H4ClN2
InChi Key:
BXXPSNZVGJRRDF-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=C1)[N+]#N
1
–
1
of
1
results
1
MP Biomedicals, Inc m-Chloroaniline Diazotate, MP Biomedicals
CAS: 17333-84-5 Molecular Formula: C6H4ClN2+ Molecular Weight (g/mol): 139.562 InChI Key: BXXPSNZVGJRRDF-UHFFFAOYSA-N Synonym: benzenediazonium, 3-chloro,c.i.azoic diazo component 2,m-chloroaniline diazotate PubChem CID: 87059 IUPAC Name: 3-chlorobenzenediazonium SMILES: C1=CC(=CC(=C1)Cl)[N+]#N