Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Chloroaniline Diazotate, MP Biomedicals

Catalog No. ICN209810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209810 Supplier MP Biomedicals, Inc Supplier No. 0520981010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17333-84-5
Molecular Formula C6H4ClN2+
Molecular Weight (g/mol) 139.562
InChI Key BXXPSNZVGJRRDF-UHFFFAOYSA-N
Synonym benzenediazonium, 3-chloro, c.i.azoic diazo component 2, m-chloroaniline diazotate
PubChem CID 87059
IUPAC Name 3-chlorobenzenediazonium
SMILES C1=CC(=CC(=C1)Cl)[N+]#N

Specifications

Quantity 10 g
Formula Weight 127.52
Chemical Name or Material m-Chloroaniline Diazotate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.