accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)(S)NH2(S)HO(S)H3CCH3OHNH3CCH3NH2OCH3H3COCH3CH3OO(S)(S)NH2(S)HO(S)H3CCH3OHNH3CCH3NH2OCH3H3COCH3OHOOHO(E)

CAS RN 173334-58-2

IUPAC Name: (2E)-but-2-enedioic acid; bis((2S,4S,5S,7S)-5-amino-N-(2-carbamoyl-2,2-dimethylethyl)-4-hydroxy-7-{[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl}-8-methyl-2-(propan-2-yl)nonanamide)
Synonyms: aliskiren hemifumarate aliskiren fumarate
Molecular Weight (g/mol): 1219.61
Molecular Formula: C64H110N6O16
InChi Key: KLRSDBSKUSSCGU-KRQUFFFQSA-N
SMILES: OC(=O)\C=C\C(O)=O.COCCCOC1=CC(C[C@@H](C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(=O)NCC(C)(C)C(N)=O)C(C)C)=CC=C1OC.COCCCOC1=CC(C[C@@H](C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(=O)NCC(C)(C)C(N)=O)C(C)C)=CC=C1OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results