accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO(E)

CAS RN 1734-79-8

IUPAC Name: (2E)-3-(4-nitrophenyl)prop-2-enal
Synonyms: 4-nitrocinnamaldehyde
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: ALGQVMMYDWQDEC-OWOJBTEDSA-N
SMILES: [O-][N+](=O)C1=CC=C(\C=C\C=O)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)