accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHHO

CAS RN 17345-66-3

IUPAC Name: 4-benzylbenzene-1,2,3-triol
Synonyms: 4-benzylbenzene-1,2,3-triol
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: UJWRVYWLRMVCIR-UHFFFAOYSA-N
SMILES: OC1=C(O)C(O)=C(CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results