Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trihydroxydiphenylmethane 98.0+%, TCI America™
SDP

Catalog No. T22595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22595G Supplier TCI America Supplier No. T22595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17345-66-3
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD08276435
InChI Key UJWRVYWLRMVCIR-UHFFFAOYSA-N
Synonym 4-Benzylpyrogallol
PubChem CID 14121594
IUPAC Name 4-benzylbenzene-1,2,3-triol
SMILES OC1=C(O)C(O)=C(CC2=CC=CC=C2)C=C1

Specifications

Melting Point 96°C
Boiling Point 255°C
Quantity 5 g
Formula Weight 216.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trihydroxydiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.