accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBr

CAS RN 17347-32-9

IUPAC Name: 6-bromo-1-benzothiophene
Synonyms: 6-bromo-1-benzothiophene
Molecular Weight (g/mol): 213.09
Molecular Formula: C8H5BrS
InChi Key: OQIMJOXSDVGEBU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CSC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)