Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromobenzo[b]thiophene 95.0+%, TCI America™
SDP

Catalog No. B55231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B55231G Supplier TCI America Supplier No. B55231G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17347-32-9
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD11226847
InChI Key OQIMJOXSDVGEBU-UHFFFAOYSA-N
PubChem CID 12744400
IUPAC Name 6-bromo-1-benzothiophene
SMILES C1=CC(=CC2=C1C=CS2)Br

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 140°C
Quantity 1 g
Formula Weight 213.09
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromobenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.