accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOO

CAS RN 17347-61-4

IUPAC Name: 3,3-dimethyloxolane-2,5-dione
Synonyms: 3,3-dimethyloxolane-2,5-dione
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: ACJPFLIEHGFXGP-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)OC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)