Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylsuccinic anhydride, 98%

Catalog No. AC308760050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308760050 5 g Glass bottle
AC308760010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308760050 Supplier Thermo Scientific Chemicals Supplier No. 308760050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17347-61-4 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17347-61-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00022608
InChI Key ACJPFLIEHGFXGP-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic anhydride, 3,3-dimethyldihydrofuran-2,5-dione, unii-tl903a7h7v, 2,5-furandione, dihydro-3,3-dimethyl, 2,2-dimethyl succinic anhydride, 2,2-dimethylsuccinicanhydride, dihydro-3,3-dimethylfuran-2,5-dione, 2,2-dimethylsuccinic acid anhydride, dihydro-3,3-dimethyl-2,5-furandione, ksc490q1j
PubChem CID 87067
IUPAC Name 3,3-dimethyloxolane-2,5-dione
SMILES CC1(C)CC(=O)OC1=O

Specifications

Melting Point 26°C to 30°C
Color White
CAS Min % 97.5
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Low Melting Solid
Assay Percent Range 98%
Boiling Point 219°C to 220°C
Formula Weight 128.13
Percent Purity 98%
Chemical Name or Material 2, 2-Dimethylsuccinic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-371-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.