CAS RN 17372-87-1
Thermo Scientific Chemicals Eosin Y, pure, Indicator grade
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2';,4';,5';,7';-Tetrabromofluorescein, disodium salt,Eosin Yellowish,2';, 4';, 5'; PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Eosin Y (Certified Biological Stain), Fisher Chemical™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: FD&C Red No. 22,Acid Red 87 PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Thermo Scientific Chemicals Eosin Y, pure, certified
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 691.85 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2',4',5',7'-Tetrabromofluorescein, disodium salt,Eosin Yellowish,C.I. 45380,2', 4', 5' PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Thermo Scientific Chemicals Eosin Y, pure, high purity, biological stain
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2',4',5',7'-Tetrabromofluorescein, disodium salt,Eosin Yellowish,2', 4', 5' PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Eosin Y Disodium Salt, 10% Solution, MP Biomedicals™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid red 87, Eosin yellowish solution, Eosin Y solution, Eosin Y PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
MilliporeSigma™ Calbiochem™ Protein Methyltransferase Inhibitor, AMI-5
The Protein Methyltransferase Inhibitor, AMI-5, also referenced under CAS 17372-87-1, controls the biological activity of Protein Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Eosin Y, Reagent, ACS, Spectrum™ Chemical
CAS: 17372-87-1 Molecular Formula: C20H6Br4Na2O5 Molecular Weight (g/mol): 691.86 InChI Key: SEACYXSIPDVVMV-UHFFFAOYSA-L IUPAC Name: disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Eosin Y Solution, Sigma-Aldrich
CAS: 17372-87-1 Molecular Weight (g/mol): 691.85 g/mol
Eosin Y Disodium Salt, Spectrum™ Chemical
CAS: 17372-87-1 Molecular Weight (g/mol): 691.85
Eosin Y TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 17372-87-1 Molecular Formula: C20H6Br4Na2O5 Molecular Weight (g/mol): 691.86 InChI Key: SEACYXSIPDVVMV-UHFFFAOYSA-L IUPAC Name: disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Eosin Y Disodium Salt, MP Biomedicals
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2',4',5',7'-Tetrabromofluorescein disodium salt,2', 4', 5',Acid Red 87 PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Thermo Scientific Chemicals Eosin Yellowish
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00133309 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: eosin yellowish,eosine,bromo acid,eosine yellowish,eosine g,bromoeosine,c.i. acid red 87,sodium eosine,sodium eosinate,eosine salt free PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Acid Red 87 90.0+%, TCI America™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: Eosine, Tetrabromofluorescein Sodium Salt PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
- 1
- 2