Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Solution, Sigma-Aldrich

Catalog No. 01259084 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-084 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-084 Supplier MilliporeSigma Supelco Supplier No. 318906500ML
Add to Cart
Add to Cart

Eosin Y, also known as Eosin yellowish is a widely used xanthene dye. It is a counterstain that is used to distinguish between various connective tissue fibers and matrices, and the cytoplasm of different types of cells. Eosin in combination with hematoxylin is useful to demonstrate the general histological architecture of a tissue.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17372-87-1
Molecular Weight (g/mol) 691.85 g/mol

Specifications

Color Red
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.