accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1738-77-8

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-4-methylpentanoate
Synonyms: benzyl (2S)-2-amino-4-methylpentanoate; para-toluene sulfonate
Molecular Weight (g/mol): 393.5
Molecular Formula: C20H27NO5S
InChi Key: QTQGHKVYLQBJLO-YDALLXLXSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)