Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Leucine benzyl ester p-toluenesulfonate, 98%, Thermo Scientific™

Catalog No. AAH6247406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6247406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6247406 Supplier Thermo Scientific Chemicals Supplier No. H6247406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-77-8
Molecular Formula C20H27NO5S
Molecular Weight (g/mol) 393.50
MDL Number MFCD00066117
InChI Key QTQGHKVYLQBJLO-YDALLXLXSA-N
Synonym l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
PubChem CID 6451312
IUPAC Name benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 156°C to 160°C
Beilstein 3566840
Quantity 5 g
Formula Weight 393.5
Percent Purity 98%
Chemical Name or Material L-Leucine benzyl ester p-toluenesulfonate
EINECSNumber 217-095-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.