accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOH2NOHNOO

CAS RN 1738-82-5

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-(2-aminoacetamido)acetate
Synonyms: benzyl 2-(2-aminoacetamido)acetate; para-toluene sulfonate
Molecular Weight (g/mol): 394.44
Molecular Formula: C18H22N2O6S
InChi Key: RWBJLSORGVEMHA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NCC(=O)NCC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results