Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycylglycine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. G04275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04275G Supplier TCI America Supplier No. G04275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1738-82-5
Molecular Formula C18H22N2O6S
Molecular Weight (g/mol) 394.44
MDL Number MFCD00050678
InChI Key RWBJLSORGVEMHA-UHFFFAOYSA-N
Synonym Diglycine Benzyl Ester p-Toluenesulfonate, H-Gly-Gly-OBzl.Tos-OH
PubChem CID 337780
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl 2-(2-aminoacetamido)acetate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.NCC(=O)NCC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 155°C
Color White
Quantity 5 g
Formula Weight 394.44
Percent Purity ≥98.0% (HPLC,N,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycylglycine Benzyl Ester p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.