accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SOOOOOOHOHOOOOHOHOOHOOHOOOHOHOOOHOHOOHOHOOOOOHHOHOOHHOHOOHHOOHOHOHH3C

CAS RN 174010-62-9

IUPAC Name: [41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecahydroxy-10,15,20,25,30,35,40-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontan-5-yl]methyl 2,4,6-trimethylbenzene-1-sulfonate
Synonyms: [41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecahydroxy-10,15,20,25,30,35,40-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontan-5-yl]methyl 2,4,6-trimethylbenzenesulfonate
Molecular Weight (g/mol): 1479.37
Molecular Formula: C57H90O42S
InChi Key: JINRRYVZBGTIER-UHFFFAOYNA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results