Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono-6-O-mesitylenesulfonyl-gamma-cyclodextrin 90.0+%, TCI America™
SDP

Catalog No. M12121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12121G Supplier TCI America Supplier No. M12121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174010-62-9
Molecular Formula C57H90O42S
Molecular Weight (g/mol) 1479.365
MDL Number MFCD00671559
InChI Key JINRRYVZBGTIER-SRQMLJPDSA-N
PubChem CID 131864313
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)OCC2C3C(C(C(O2)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(C(C9O)O)OC1C(OC(O3)C(C1O)O)CO)CO)CO)CO)CO)CO)CO)O)O)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 1479.37
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mono-6-O-mesitylenesulfonyl-gamma-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.