accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17403-09-7

IUPAC Name: 4-(1H-indol-3-yl)piperidin-1-ium
Synonyms: 4-(1H-indol-3-yl)piperidin-1-ium
Molecular Weight (g/mol): 201.29
Molecular Formula: C13H17N2
InChi Key: KAIRZPVWWIMPFT-UHFFFAOYSA-O
SMILES: C1CC(CC[NH2+]1)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results