accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOCH3

CAS RN 1743-60-8

IUPAC Name: 7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: 7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H-cyclopenta[a]phenanthren-1-yl acetate
Molecular Weight (g/mol): 314.43
Molecular Formula: C20H26O3
InChi Key: QAHOQNJVHDHYRN-UHFFFAOYNA-N
SMILES: CC(=O)OC1CCC2C3CCC4=C(C=CC(O)=C4)C3CCC12C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results