Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Estradiol-17-acetate, MP Biomedicals™

Catalog No. ICN15794391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15794391 1 g
ICN15794383 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15794391 Supplier MP Biomedicals, Inc Supplier No. 0215794391

Chemical Identifiers

CAS 1743-60-8
Molecular Formula C20H26O3
Molecular Weight (g/mol) 314.43
MDL Number MFCD00056537
InChI Key QAHOQNJVHDHYRN-UHFFFAOYNA-N
Synonym beta-estradiol 17-acetate, 17beta-acetylestradiol, unii-2vm9ho33ru, estradiol-17o-acetate, 2vm9ho33ru, 17beta-estradiol 17-acetate, estra-1,3,5 10-triene-3,17beta-diol 17-acetate, estradiol 17-acetate, estradiol 17-monoacetate
PubChem CID 6852404
ChEBI CHEBI:79735
IUPAC Name 7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
SMILES CC(=O)OC1CCC2C3CCC4=C(C=CC(O)=C4)C3CCC12C

Specifications

Quantity 1 g
Formula Weight 314.4
Physical Form Crystalline
Chemical Name or Material β-Estradiol-17-Acetate
storageNote1 Protect from light.
Recommended Storage Store at room temperature (15° to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.