accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 174350-08-4

CAS RN 174350-08-4

IUPAC Name: 4-(2,3-difluoro-4-methylphenyl)-4'-ethyl-1,1'-bi(cyclohexane)
Synonyms: 4-(2,3-difluoro-4-methylphenyl)-4'-ethyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 320.47
Molecular Formula: C21H30F2
InChi Key: GDXLAJNVPUQWRF-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(CC1)C1=C(F)C(F)=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results