accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNS

CAS RN 17452-14-1

IUPAC Name: 1-(4-methoxyphenyl)-2,3-dihydro-1H-imidazole-2-thione
Synonyms: 1-(4-methoxyphenyl)-3H-imidazole-2-thione
Molecular Weight (g/mol): 206.26
Molecular Formula: C10H10N2OS
InChi Key: LIEBCNQPMNARMP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1C=CNC1=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results