accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1746-11-8

IUPAC Name: 2-methyl-2,3-dihydro-1-benzofuran
Synonyms: 2-methyl-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: BWCJVGMZEQDOMY-UHFFFAOYNA-N
SMILES: CC1CC2=CC=CC=C2O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results