accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)OOFFF

CAS RN 17463-43-3

IUPAC Name: (2R)-2-azaniumyl-3,3,3-trifluoropropanoate
Synonyms: (2R)-2-ammonio-3,3,3-trifluoropropanoate
Molecular Weight (g/mol): 143.07
Molecular Formula: C3H4F3NO2
InChi Key: HMJQKIDUCWWIBW-PVQJCKRUSA-N
SMILES: [NH3+][C@H](C([O-])=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)