accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHNF

CAS RN 174669-73-9

IUPAC Name: (2-fluoropyridin-3-yl)boronic acid
Synonyms: 2-fluoropyridin-3-ylboronic acid
Molecular Weight (g/mol): 140.91
Molecular Formula: C5H5BFNO2
InChi Key: YUHZIUAREWNXJT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CN=C1F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results