Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F07395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07395G 5 g
F0739-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07395G Supplier TCI America Supplier No. F07395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 174669-73-9 | C5H5BFNO2 | 140.91 g/mol

Specifications

Chemical Name or Material 2-Fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 174669-73-9
Melting Point 172°C
Color White-Yellow
Molecular Formula C5H5BFNO2
Quantity 5 g
MDL Number MFCD03411557
Synonym 2-fluoropyridine-3-boronic acid, 2-fluoro-3-pyridylboronic acid, 2-fluoropyridin-3-yl boronic acid, 2-fluoro-3-pyridineboronic acid, 2-fluoropyridin-3-yl-3-boronic acid, 2-fluoro-3-pyridilboronic acid, 2-fluoropryridine-3-boronic acid, boronic acid, 2-fluoro-3-pyridinyl, 2-fluoro-3-pyridyl boronic acid, 2-fluoropyridine-3-boronicacid
InChI Key YUHZIUAREWNXJT-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CN=C1F
IUPAC Name (2-fluoropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 140.91
PubChem CID 2783270
Formula Weight 140.91
Percent Purity 98.0 to 115.0%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.