accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1747-60-0

IUPAC Name: 6-methoxy-1,3-benzothiazol-2-amine
Synonyms: 2-amino-6-methoxybenzothiazole
Molecular Weight (g/mol): 180.23
Molecular Formula: C8H8N2OS
InChi Key: KZHGPDSVHSDCMX-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(N)SC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)