accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCOOCHOOCHH3COOHCOCH3HfCH3H3CH3CCH3CH3H3C

CAS RN 17475-67-1

IUPAC Name: hafnium(4+) tetrakis(2,4-dioxopentan-3-ide)
Synonyms: hafnium(4+) tetraacetylacetonate
Molecular Weight (g/mol): 574.92
Molecular Formula: C20H28HfO8
InChi Key: GXNMJQUZCICHBX-UHFFFAOYSA-N
SMILES: [Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results