Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) 2,4-pentanedionate, 97%

Catalog No. AA4246314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4246314 25 g
AA4246303 1 g
AA4246306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4246314 Supplier Thermo Scientific Chemicals Supplier No. 04246314
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17475-67-1 | C20H28HfO8 | 574.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17475-67-1
Molecular Formula C20H28HfO8
Molecular Weight (g/mol) 574.92
MDL Number MFCD00044997
InChI Key GXNMJQUZCICHBX-UHFFFAOYSA-N
Synonym hafnium iv acetylacetonate, hafnium-2,4-pentanedionate, hafnium; z-4-hydroxypent-3-en-2-one, 3z-4-tris 2z-4-oxopent-2-en-2-yl oxy hafnio oxy pent-3-en-2-one
PubChem CID 11330624
IUPAC Name hafnium;(Z)-4-hydroxypent-3-en-2-one
SMILES [Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 193°C
Odor Characteristic
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 574.93
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Hafnium (IV) 2,4-pentanedionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.