accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 174899-65-1

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium trifluoroacetate
Synonyms: 1-ethyl-3-methylimidazolium trifluoroacetate
Molecular Weight (g/mol): 224.18
Molecular Formula: C8H11F3N2O2
InChi Key: JOKVYNJKBRLDAT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(F)(F)F.CCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results