accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CH3CFFFSOOO

CAS RN 174899-72-0

IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 3-ethyl-1,2-dimethylimidazol-1-ium triflate
Molecular Weight (g/mol): 274.26
Molecular Formula: C8H13F3N2O3S
InChi Key: HMAHVRVKBSHMJX-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCN1C=C[N+](C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results