Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 98%, Thermo Scientific™

Catalog No. AAH5972606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5972606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5972606 Supplier Thermo Scientific Chemicals Supplier No. H5972606

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174899-72-0
Molecular Formula C8H13F3N2O3S
Molecular Weight (g/mol) 274.258
MDL Number MFCD07784438
InChI Key HMAHVRVKBSHMJX-UHFFFAOYSA-M
Synonym 1-ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-ethyl-2,3-dimethylimidazolium triflate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium trifluoromethanesulfonate, 3-ethyl-1,2-dimethylimidazol-1-ium triflate, dsstox_cid_29089, dsstox_rid_83308, dsstox_gsid_49233, 1-ethyl-2 3-dimethylimidazolium trifluor, 1-ethyl-2,3-dimethylimidazol-1-ium triflate
PubChem CID 16210995
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 274.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.