CAS RN 174899-94-6
IUPAC Name:
1-butyl-3-methyl-1H-imidazol-3-ium trifluoroacetate
Synonyms:
1-butyl-3-methylimidazolium trifluoroacetate
Molecular Weight (g/mol):
252.24
Molecular Formula:
C10H15F3N2O2
InChi Key:
QPDGLRRWSBZCHP-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C(F)(F)F.CCCCN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1-Butyl-3-methylimidazolium Trifluoroacetate 97.0+%, TCI America™
CAS: 174899-94-6 Molecular Formula: C10H15F3N2O2 Molecular Weight (g/mol): 252.237 MDL Number: MFCD08458921 InChI Key: QPDGLRRWSBZCHP-UHFFFAOYSA-M PubChem CID: 10125280 IUPAC Name: 1-butyl-3-methylimidazol-3-ium;2,2,2-trifluoroacetate SMILES: CCCCN1C=C[N+](=C1)C.C(=O)(C(F)(F)F)[O-]