accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCOOCHOOCHH3COOHCOCH3ZrCH3H3CCF3CF3CF3CF3

CAS RN 17499-68-2

IUPAC Name: zirconium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide)
Synonyms: zirconium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 703.55
Molecular Formula: C20H16F12O8Zr
InChi Key: QOUVICDMOZRTCK-UHFFFAOYSA-N
SMILES: [Zr+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results