Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) 98.0+%, TCI America™
SDP

Catalog No. T07471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07471G Supplier TCI America Supplier No. T07471G

Chemical Identifiers

CAS 17499-68-2
Molecular Formula C20H16F12O8Zr
Molecular Weight (g/mol) 703.55
MDL Number MFCD00015317
InChI Key QOUVICDMOZRTCK-UHFFFAOYSA-N
Synonym zirconium iv tetrakis 1,1,1-trifluoro-4-oxo-2-pentene-2-olate
PubChem CID 101644371
IUPAC Name zirconium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide)
SMILES [Zr+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 703.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.