accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-63-4

IUPAC Name: 2-(2,4-difluorophenoxy)pyridin-3-amine
Synonyms: 2-(2,4-difluorophenoxy)pyridin-3-amine
Molecular Weight (g/mol): 222.2
Molecular Formula: C11H8F2N2O
InChi Key: LCPVQAHEFVXVKT-UHFFFAOYSA-N
SMILES: NC1=C(OC2=C(F)C=C(F)C=C2)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results