Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Difluorophenoxy)pyridin-3-amine, 97%, Thermo Scientific™

Catalog No. BTB08015DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BTB08015DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB08015DA Supplier Thermo Scientific Chemicals Supplier No. BTB08015DA

missing translation for 'validMainframeMsg'

Specifications

CAS 175135-63-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H8F2N2O
Quantity 1g
MDL Number MFCD00067802
Synonym 2-2,4-difluorophenoxy pyridin-3-amine, 3-amino-2-2,4-difluorophenoxy pyridine, 3-pyridinamine,2-2,4-difluorophenoxy, 2-2,4-difluorophenoxy-3-pyridylamine, zlchem 686, pubchem2980, maybridge1_001738, 2-2,4-difluorophenoxy-3-pyridinamine, 2-2,4-difluorophenoxy pyridine-3-amine, 3-pyridinamine, 2-2,4-difluorophenoxy
InChI Key LCPVQAHEFVXVKT-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)OC2=C(C=C(C=C2)F)F)N
IUPAC Name 2-(2,4-difluorophenoxy)pyridin-3-amine
Molecular Weight (g/mol) 222.195
PubChem CID 2735906
Formula Weight 222.19
Percent Purity 97%
Chemical Name or Material 2-(2, 4-Difluorophenoxy)pyridin-3-amine
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.