accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 175136-12-6

IUPAC Name: 7-methyl-2,3-dihydro-1H-inden-4-yl acetate
Synonyms: 7-methyl-2,3-dihydro-1H-inden-4-yl acetate
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: MABWFAYFFNHAOF-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C)C2=C1CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results