Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methyl-2,3-dihydro-1H-inden-4-yl acetate, 97%, Thermo Scientific™

Catalog No. BTB09402DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09402DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09402DA Supplier Thermo Scientific Chemicals Supplier No. BTB09402DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175136-12-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00173682
InChI Key MABWFAYFFNHAOF-UHFFFAOYSA-N
Synonym 4-acetoxy-7-methylindane, acmc-1c5pj, 7-methylindan-4-yl acetate, 7-methyl-2,3-dihydro-1h-inden-4-yl acetate, 1h-inden-4-ol,2,3-dihydro-7-methyl-,4-acetate, 1h-inden-4-ol,2,3-dihydro-7-methyl-, 4-acetate
PubChem CID 2775024
IUPAC Name (7-methyl-2,3-dihydro-1H-inden-4-yl) acetate
SMILES CC(=O)OC1=CC=C(C)C2=C1CCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 7-Methyl-2, 3-dihydro-1H-inden-4-yl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.