accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrON

CAS RN 175136-65-9

IUPAC Name: 3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
Synonyms: 3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 254.08
Molecular Formula: C10H8BrNO2
InChi Key: NVZISNDFOXXQSQ-UHFFFAOYSA-N
SMILES: COC1=CC(C(=O)CC#N)=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results