Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile, Thermo Scientific™

Catalog No. CD05258EB
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD05258EB 25 g
CD05258DA 1 g
CD05258EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD05258EB Supplier Thermo Scientific Chemicals Supplier No. CD05258EB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175136-65-9
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
InChI Key NVZISNDFOXXQSQ-UHFFFAOYSA-N
Synonym 3-2-bromo-5-methoxyphenyl-3-oxopropanenitrile, maybridge1_002847, 3-2-bromo-5-methoxy-phenyl-3-oxo-propanenitrile
PubChem CID 2804251
IUPAC Name 3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
SMILES COC1=CC(=C(C=C1)Br)C(=O)CC#N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 254.08
Percent Purity 97%
Chemical Name or Material 3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.