accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175137-46-9

IUPAC Name: 5-cyclopropyl-1H-pyrazol-3-amine
Synonyms: 5-cyclopropyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: MXVAGCQKBDMKPG-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1CC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results